missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Accessories for Biosart™ 100 Membrane Filtration
Biosart™ 100 Membrane Filtration is designed specifically for microbiological testing of pharmaceuticals, food, beverages, water and other liquids.
11.35 € - 204.00 €
Specifications
Disposable | Reusable |
---|---|
For Use With (Equipment) | Sartorius Biosart 100 |
Sterility | Non-sterile |
Product Code | Brand | Type | Material | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Material | For Use With (Application) | Price | Quantity & Availability | ||||
11765545
|
Sartorius
16417 |
Adapter | ABS Copolymer | For easy transfer of the membrane onto agar |
11.35 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||
11392120
|
Sartorius
16414 |
Adapter | Silicone | Fits Biosart 100 Monitor onto 16840 or 16841 base |
15.10 €
Each |
In Stock
Log in to see stock. |
||||
10730403
|
Sartorius
16835 |
Adapter | Stainless Steel | Fits Biosart 100 Monitors onto Combisart manifolds 16844, 16842 and 16843 |
204.00 €
Each |
In Stock
Log in to see stock. |
||||
Description
Accessories Descriptions
Adapter, ABS copolymer - 14-555-780- Biosart 100 Membrane Lifter
- For easy transfer of the membrane onto agar
- Biosart 100 Adapter
- Fits Biosart 100 Monitor onto 50 mm supports
- Biosart 100 Adapter
- 50 mm stainless-steel frit and silicone gasket
- Fits Biosart 100 Monitor onto 16840 or 16841 base
- Biosart 100 Adapter to accommodate various funnel types
- Fits Biosart 100 Monitors onto Combisart™ manifolds 16844, 16842 and 16843
- Replacement stopper for 16385
- 16385 adapts Biosart 100 Monitors on Combisart manifold
Spécification
Reusable | |
Sartorius Biosart 100 | |
Non-sterile |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Sartorius Accessories for Biosart™ 100 Membrane Filtration