Learn More
Sartorius Accessories for Biosart™ 100 Membrane Filtration
Biosart™ 100 Membrane Filtration is designed specifically for microbiological testing of pharmaceuticals, food, beverages, water and other liquids.
Brand:  Sartorius 16414
11.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11392120
  • 15.10 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Accessories Descriptions
Adapter, ABS copolymer - 14-555-780
  • Biosart 100 Membrane Lifter
  • For easy transfer of the membrane onto agar
Adapter, Polypropylene - 14-555-779
  • Biosart 100 Adapter
  • Fits Biosart 100 Monitor onto 50 mm supports
Adapter, Silicone - 14-555-778
  • Biosart 100 Adapter
  • 50 mm stainless-steel frit and silicone gasket
  • Fits Biosart 100 Monitor onto 16840 or 16841 base
Adapter, Stainless Steel - 14-555-781
  • Biosart 100 Adapter to accommodate various funnel types
  • Fits Biosart 100 Monitors onto Combisart™ manifolds 16844, 16842 and 16843
Stopper, Silicone - 14-555-817
  • Replacement stopper for 16385
  • 16385 adapts Biosart 100 Monitors on Combisart manifold
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Reusable
Sartorius Biosart 100
Non-sterile
Adapter
Manifold
Silicone
Fits Biosart 100 Monitor onto 16840 or 16841 base
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius Accessories for Biosart™ 100 Membrane Filtration > Biosart 100 Membrane Filtration Accessory; Type: Adapter; Material: Silicone; Application: Fits Biosart 100 Monitor onto 16840 or 16841 base