missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Glass Filtration Holder
This glass vacuum filtration holder has been designed for particle testing, clarification and sterile filtration with the use of 24 or 25 mm filters.
Brand: Sartorius 16306
201.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
The device is delivered with a glass frit filter support that ensures a uniform distribution of the retained particles. Therefore it is recommended when the residue on the filter surface is of interest.Specifications
Glass | |
Each | |
30 mL |
3 cm2 | |
Vacuum Filter Holder | |
2.5 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction