Learn More
Sartorius Nutrient Pad Sets (NPS) 47mm diameter
Sterile, dehydrated culture media which is individually inserted in a petri dish and sterilized
251.00 € - 268.00 €
Specifications
Form | Dehydrated |
---|---|
Manufacturing Origin | Germany |
Sterility | Sterile |
Test Type | Microbial Detection |
Packaging Type | Individually Packaged |
Product Code | Brand | Color | Grid Color | Pore Size | Media Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Grid Color | Pore Size | Media Type | Price | Quantity & Availability | |||||
11760676
|
Sartorius
14090--47------N |
Green | Green | 0.45μm | Yeast Extract |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10337591
|
Sartorius
14051--47------N |
Green | Dark Green | 0.45μm | Azide |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10737622
|
Sartorius
14053--47------N |
White | Green | 0.45μm | Endo |
268.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10357591
|
Sartorius
14055--47------N |
Green | Dark Green | 0.45μm | Standard TTC |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10265461
|
Sartorius
14056--47------N |
White | Green | 0.45μm | Tergitol TTC |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10052141
|
Sartorius
14057--47------N |
Green | Dark Green | 0.45μm | Bismuth Sulfite | N/A | ||||||
10104181
|
Sartorius
14058--47------N |
Gray | White | 0.65μm | Bismuth Sulfite |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10357301
|
Sartorius
14059--47------N |
White | Green | 0.45μm | VLB-S7-S |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10072141
|
Sartorius
14061--47------N |
Gray | White | 0.65μm | Lysine | N/A | ||||||
10402002
|
Sartorius
14062--47------N |
Green | Dark Green | 0.45μm | Orange Serum |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10012371
|
Sartorius
14063--47------N |
Green | Dark Green | 0.45μm | Caso |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10799201
|
Sartorius
14064--47------N |
Green | Dark Green | 0.45μm | Standard |
265.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10235751
|
Sartorius
14065--47------N |
Green | Dark Green | 0.45μm | Weman | N/A | ||||||
10205801
|
Sartorius
14066--47------N |
White | Green | 0.45μm | Glucose Tryptone |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10367401
|
Sartorius
14068--47------N |
White | Green | 0.45μm | m-FC |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10747622
|
Sartorius
14067--47------N |
White | Green | 0.45μm | Lauryl Sulphate | N/A | ||||||
10707052
|
Sartorius
14069--47------N |
Gray | White | 0.65μm | Sabouraud Agar |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10662412
|
Sartorius
14070--47------N |
White | Green | 0.65μm | Schaufus Pottinger |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10357641
|
Sartorius
14075--47------N |
White | Green | 0.45μm | Cetrimide |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10490462
|
Sartorius
14074--47------N |
White | Green | 0.45μm | Chapman |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10317741
|
Sartorius
14072--47------N |
White | Green | 1.2μm | Schaufus Pottinger | N/A | ||||||
11780656
|
Sartorius
14076--47------N |
Green | Dark Green | 0.45μm | TGE |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10377211
|
Sartorius
14079--47------N |
Green | Dark Green | 0.45μm | Jus de Tomate |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10215611
|
Sartorius
14082--47------N |
White | Green | 0.45μm | ECD | N/A | ||||||
11760666
|
Sartorius
14083--47------N |
Gray | White | 0.65μm | Schaufus Pottinger |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10515331
|
Sartorius
14084--47------N |
Green | Dark Green | 0.45μm | R2A |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10172222
|
Sartorius
14080--47------N |
Gray | White | 0.8μm | Schaufus Pottinger |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10012351
|
Sartorius
14086--47------N |
Gray | White | 0.8μm | Malt Extract |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15942500
|
Sartorius
14086--47----CCN |
White | Black | 0.45μm | Malt Extract |
251.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11740676
|
Sartorius
14089--47------N |
Green | Dark Green | 0.45μm | Wallerstein |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10235851
|
Sartorius
14087--47------N |
Gray | White | 0.45μm | Chromocult |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10295891
|
Sartorius
14097--47------N |
Gray | White | 0.45μm | MacConkey |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11710686
|
Sartorius
14096--47------N |
Gray | White | 0.45μm | MacConkey |
254.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Sartorius Nutrient Pad Sets are used in the membrane filter methods. The membrane filter method allows large sample volumes with low microbial count to be tested. Inhibitors, such as essential oils or disinfectants, can be rinsed out after filtration, before incubation.Practical and easy to handle, Nutrient Pads reduce labor and simplify many microbiological testing procedures. Microorganisms require the appropriate culture media for optimal growth.Nutrient Pad Sets are sterile, dehydrated culture media. Once they are moistened with 3.0 - 3.5ml of sterile and demineralized (or destilled) water they are ready-to-use immediately. The level of moisture is optimal when an excess ring of water surrounding the NPS is visible.All Nutrient Pad Set types are supplied with the appropriate membrane filters, which are also individually, sterile packaged. The membrane filters tailored to meet the special requirements of microbial detection are available with 47 or 50mm diameter. The standard NPS box contains 100 sterile Nutrient Pads, each of which is individually inserted in a petri dish and sterilized. Ten each of these petri dishes are sealed in an aluminum bag. This special packaging in bags protects the sensitive formula constituents of the Nutrient Pad during transport and storage from fluctuations in humidity and temperature. As a result, it guarantees the high quality of NPS throughout their entire shelf life up to 24 months. More than 30 different Nutrient Pad Set types are available to meet the diverse objectives of microbiological analysis. Beyond the European drinking water directive, they comply with other international regulations and recommendations: international pharmacopoeias, DIN and ISO standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA, etc.
- trouble-free storage up to 24 month
- economical by time- and cost-saving
- according to international standards and references
- easy handling
- validation guide available
- consistent quality
- highly versatile
Spezifikation
Dehydrated | |
Sterile | |
Individually Packaged | |
4.7 cm |
Germany | |
Microbial Detection | |
1.85 in. |
Sicherheit und Handhabung
missing translation for 'shelfLife' : 2 Years
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.