missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 100 Disposable Ultrafiltration Units
BIOPHARMA

Vivaspin™ 100 has the highest capacity of any centrifugal ultrafiltration device, for easy and convenient handling of larger sample volumes.

Brand:  Sartorius VC1032

463.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10757822

  • 605.00 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For the concentration of samples up to 98 mL
    • Can be used either in a centrifuge or for pressure filtration
  • Combines the ease of use of centrifugal devices with the flexibility and control provided by pressurized ultrafiltration cells
  • Easy to assemble
    • Requires no tubing connections or stirring mechanisms
    • Can be adapted to equipment availability or to specific user preferences
  • For convenience, the units are simply spun in a large capacity centrifuge (carriers accepting 250 mL bottles)
  • For better process control, gas pressure can be combined with gentle orbital shaking
  • For highest simplicity of use with minimum equipment requirements, the device is pressurized and then left standing on a bench top or in a refrigerator
  • The concentrator features a vertical membrane that minimizes fouling, while the built-in deadstop of 350 μL will prevent concentration to dryness.

Features:

  • Easy to handle
  • High flux PES membrane
  • Vertical twin membrane for ultra-fast concentration
  • High retentate recovery (90%)
  • Total process flexibility
  • Compatible with swing out rotors
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
PES
20 to 98 mL
10 Pk.
Centrifugal Ultrafiltration
50 kDa
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.