Learn More
Sartorius Vivaspin™ 100 Disposable Ultrafiltration Units
Vivaspin™ 100 has the highest capacity of any centrifugal ultrafiltration device, for easy and convenient handling of larger sample volumes.
135.00 € - 605.00 €
Specifications
Disposable | Single-use |
---|---|
Filter Type | Ultrafiltration |
Material (Membrane) | PES |
Sterility | Non-sterile |
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | ||||
10489321
|
Sartorius
VC1001 |
10 kDa | 2 Pk. |
135.00 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10751733
|
Sartorius
VC1002 |
10 kDa | 10 Pk. |
605.00 €
Pack of 10 |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10275901
|
Sartorius
VC1022 |
30 kDa | 10 Pk. |
605.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
10066861
|
Sartorius
VC1012 |
5 kDa | 10 Pk. |
605.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
10154141
|
Sartorius
VC1021 |
30 kDa | 2 Pk. |
135.00 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10194091
|
Sartorius
VC1011 |
5 kDa | 2 Pk. |
135.00 €
Pack of 2 |
Estimated Shipment: 18-03-2025
Log in to see stock. |
||||
10757632
|
Sartorius
VC1031 |
50 kDa | 2 Pk. |
135.00 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10757822
|
Sartorius
VC1032 |
50 kDa | 10 Pk. |
605.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
10409711
|
Sartorius
VC1041 |
100 kDa | 2 Pk. |
135.00 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10552692
|
Sartorius
VC1042 |
100 kDa | 10 Pk. |
605.00 €
Pack of 10 |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10194281
|
Sartorius
VC1051 |
300 kDa | 2 Pk. |
135.00 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10066871
|
Sartorius
VC1052 |
300 kDa | 10 Pk. |
605.00 €
Pack of 10 |
Estimated Shipment: 18-03-2025
Log in to see stock. |
||||
10076871
|
Sartorius
VC1071 |
0.2 μm | 2 Pk. | N/A | |||||
10302222
|
Sartorius
VC1062 |
1000 kDa | 10 Pk. |
605.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
10290242
|
Sartorius
VC1072 |
0.2 μm | 10 Pk. |
605.00 €
Pack of 10 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||
10619161
|
Sartorius
VC1061 |
1000 kDa | 2 Pk. |
135.00 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
Description
- For the concentration of samples up to 98 mL
- Can be used either in a centrifuge or for pressure filtration
- Combines the ease of use of centrifugal devices with the flexibility and control provided by pressurized ultrafiltration cells
- Easy to assemble
- Requires no tubing connections or stirring mechanisms
- Can be adapted to equipment availability or to specific user preferences
- For convenience, the units are simply spun in a large capacity centrifuge (carriers accepting 250 mL bottles)
- For better process control, gas pressure can be combined with gentle orbital shaking
- For highest simplicity of use with minimum equipment requirements, the device is pressurized and then left standing on a bench top or in a refrigerator
- The concentrator features a vertical membrane that minimizes fouling, while the built-in deadstop of 350 μL will prevent concentration to dryness.
Features:
- Easy to handle
- High flux PES membrane
- Vertical twin membrane for ultra-fast concentration
- High retentate recovery (90%)
- Total process flexibility
- Compatible with swing out rotors
Specifications
Single-use | |
Ultrafiltration | |
PES | |
Non-sterile |