missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivaspin™ 6 Centrifugal Concentrators
Achieve fast sample concentration and high recovery. Sartorius Vivaspin™ 6 Centrifugal Concentrators are the only device of its size that can process 6mL samples. They feature twin vertical membranes and built-in dead stop pockets. Available with PES membranes in a wide range of molecular weight cut-offs.
Brand: Sartorius VS0642
954.75 EUR valid until 2024-12-31
Use promo code "21930" to get your promotional price.
This item is not returnable.
View return policy
Description
Developed to offer increased volume flexibility and performance- Compact tubes can process up to an impressive 6mL initial volume
- Optimally suited for the concentration of antibody, protein, nanoparticle and virus samples
- Universal rotor compatibility:
- Use in either swing bucket or fixed angle rotors accepting standard 15mL conical bottom test tubes
- Features premium, high-throughput PES membranes
- Twin vertical membrane design lead to unparalleled filtration speed and over 100-fold concentration
- The dead stop pocket prevents concentration to dryness for safe sample concentration
- Nearly complete concentrate recovery of over 90%
- Remaining volume is easy to see with the printed graduations and clear housing
- Modified dead stop pocket further simplifies direct pipette recovery of the final concentrate
Specifications
PES | |
2.58 cm2 | |
100/Pk. | |
122 mm |
100 kDa | |
Polycarbonate | |
6 mL |