missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 6 Centrifugal Concentrators
BIOPHARMA

Achieve fast sample concentration and high recovery. Sartorius Vivaspin™ 6 Centrifugal Concentrators are the only device of its size that can process 6mL samples. They feature twin vertical membranes and built-in dead stop pockets. Available with PES membranes in a wide range of molecular weight cut-offs.

207.00 € - 1035.00 €

Specifications

Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) PES
Sample Volume 6 mL
View More Specs

Products 20
Product Code Brand MWCO Quantity Price Quantity & Availability  
Product Code Brand MWCO Quantity Price Quantity & Availability  
16656752
View Documents
Sartorius
VS0601
10 kDa 25/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16676752
missing translation for 'viewDocuments'
Sartorius
VS0602
10 kDa 100/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10421243
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0602
10 kDa 100/Pk.
1005.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
10767461
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0611
5 kDa 25/Pk.
332.00 €
Pack of 25
In Stock
to see stock.
Add to basket
 
10293471
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0612
5 kDa 100/Pk.
1035.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
10029460
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0621
30 kDa 25/Pk.
348.00 €
Pack of 25
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
 
10384981
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0622
30 kDa 100/Pk.
1005.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
10395021
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0631
50 kDa 25/Pk.
332.00 €
Pack of 25
In Stock
to see stock.
Edge
Add to basket
 
10588191
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0632
50 kDa 100/Pk.
1005.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
10120901
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0642
100 kDa 100/Pk.
1005.00 €
Pack of 100
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
 
10009640
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0641
100 kDa 25/Pk.
348.00 €
Pack of 25
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
 
11741826
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0651
300 kDa 25/Pk.
332.00 €
Pack of 25
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
 
11740746
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0652
300 kDa 100/Pk.
1005.00 €
Pack of 100
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
 
10777461
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0661
1000 kDa 25/Pk.
348.00 €
Pack of 25
In Stock
to see stock.
Add to basket
 
10110951
missing translation for 'viewDocuments'
Sartorius
VS0662
1000 kDa 100/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10059520
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0671
0.2 μm 25/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10449512
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0672
0.2 μm 100/Pk.
1005.00 €
Pack of 100
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
 
11770746
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0692
3 kDa 100/Pk.
1005.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
10150203
BIOPHARMA
missing translation for 'viewDocuments'
Sartorius
VS0691
3 kDa 25/Pk.
332.00 €
Pack of 25
In Stock
to see stock.
Add to basket
 
10738421
missing translation for 'viewDocuments'
Sartorius
VS06S1
5, 10, 30, 50, and 100 kDa 25/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Developed to offer increased volume flexibility and performance
  • Compact tubes can process up to an impressive 6mL initial volume
  • Optimally suited for the concentration of antibody, protein, nanoparticle and virus samples
  • Universal rotor compatibility:
    • Use in either swing bucket or fixed angle rotors accepting standard 15mL conical bottom test tubes
Fast, safe and easy concentration
  • Features premium, high-throughput PES membranes
  • Twin vertical membrane design lead to unparalleled filtration speed and over 100-fold concentration
  • The dead stop pocket prevents concentration to dryness for safe sample concentration
  • Nearly complete concentrate recovery of over 90%
  • Remaining volume is easy to see with the printed graduations and clear housing
  • Modified dead stop pocket further simplifies direct pipette recovery of the final concentrate
Specifications

Specifications

Single-use
Centrifuge
6 mL
2.58 cm2
6 mL
Ultrafiltration
PES
Non-sterile
Polycarbonate
122 mm
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.