missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 6 Centrifugal Concentrators

Achieve fast sample concentration and high recovery. Sartorius Vivaspin™ 6 Centrifugal Concentrators are the only device of its size that can process 6mL samples. They feature twin vertical membranes and built-in dead stop pockets. Available with PES membranes in a wide range of molecular weight cut-offs.

Brand:  Sartorius VS0602

 View more versions of this product

Product Code. 16676752

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Developed to offer increased volume flexibility and performance
  • Compact tubes can process up to an impressive 6mL initial volume
  • Optimally suited for the concentration of antibody, protein, nanoparticle and virus samples
  • Universal rotor compatibility:
    • Use in either swing bucket or fixed angle rotors accepting standard 15mL conical bottom test tubes
Fast, safe and easy concentration
  • Features premium, high-throughput PES membranes
  • Twin vertical membrane design lead to unparalleled filtration speed and over 100-fold concentration
  • The dead stop pocket prevents concentration to dryness for safe sample concentration
  • Nearly complete concentrate recovery of over 90%
  • Remaining volume is easy to see with the printed graduations and clear housing
  • Modified dead stop pocket further simplifies direct pipette recovery of the final concentrate
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PES
2.58 cm2
100/Pk.
122 mm
10 kDa
Polycarbonate
6 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers