Learn More
Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors
Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
838.17 € - 1678.35 €
Specifications
Type | Concentrator Rotor |
---|---|
For Use With (Equipment) | DNA130 |
Rotor System | Traditional |
Product Code | Brand | Holds | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | ||||||
11718609
|
Thermo Scientific™
RD24 |
24 |
838.17 €
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
11768599
|
Thermo Scientific™
RD2MP |
2 Shallow-well Plates |
1678.35 €
Each |
In Stock
Log in to see stock. |
||||||
11778599
|
Thermo Scientific™
RD36 |
36 |
1004.00 €
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
11728609
|
Thermo Scientific™
RD48 |
48 |
841.19 €
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
11748609
|
Thermo Scientific™
RD72 |
72 |
1075.35 €
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
Description
Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
Easy to use circular rotors with different tube hole sizes to accommodate different sample sizes. These rotors require no additional accessories and fit directly into your concentrator.Specifications
Concentrator Rotor | |
DNA130 | |
Traditional |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors