Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Select from one of these rotors to fit the type of sample volume and various sample formats for your specific application. Choose from various size of tubes, flasks, mircrowell plates.
Brand:  Thermo Scientific™ RH24-18
 View more versions of this product
Product Code. 11798589
  • 1165.80 € / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
SpeedVac Rotors for Medium Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into your concentrator
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24-place; 18 x 52mm mini scint vials
SPD1030, SPD120, SPD130, SPD140DDA
RH24-18
Rotor
18 x 52 mm Mini-scintillation
Traditional