missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ SC-200 Series Sample Concentrators

Quickly concentrate multiple samples in a block at once.

Brand:  Cole-Parmer™ 36620-42

1139.32 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 17986082

  • 1475.00 € / 1 unit
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Variable needle position suits various Dri-Block™ inserts
  • Small size makes it ideal for use under a fume hood
  • Needles, block, and block heater sold separately
  • Concentrator blows an inert gas over the surface of your samples via stainless steel needles (not included).
  • Blown gas in combination with the heat from the heater produce ideal conditions for fast efficient evaporation.
  • Insert needles into a silicone membrane in any configuration.
  • Fully adjustable gas reservoir provides accurate height control.
  • Maximum vertical traveling distance is 32 cm.
  • Maximum gas usage is 15 L/min.
  • Order PTFE-coated needles for use with corrosive samples.
Specifications

Specifications

SC-200
For Tubes
Dri-Block™ Inserts
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ SC-200 Series Sample Concentrators > Description: For Tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.