Learn More
GPE Scientific™ Schlenk Flask, Cylindrical with Glass Tap and Cone
GPE Scientific™ Schlenk Flask, Cylindrical with Glass Tap and Cone are specially designed for use with Schlenk lines and vacuum manifolds and are available in different capacities.
Brand:  GPE Scientific™ SCG/100
 View more versions of this product
Product Code. 15814884
  • 88.20 € / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cylindrical, Schlenk-style flask
  • Includes glass tap and cone
  • Available in different capacities
  • Available with different joint sizes
Specifications
Specifications
100 mL
Glass tap and cone
Cylindrical
Cylindrical
Schlenk lines and vacuum manifolds
Cylindrical Schlenk flask with tap and cone
Joint size: 24