Learn More
SI Analytics™ ScienceLine Ion Selective Electrode
SI Analytics ScienceLine Ion Selective Electrodes possess the advantage of a much lower temperature sensitivity and a metal ion free reference system.
Brand:  SI Analytics™ 285130430
 View more versions of this product
Product Code. 10405253
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • ScienceLine sensors for ammonia, sodium, oxygen, ion-selective indicator electrodes
  • Also measure pH and temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Copper
Cu 60
0°C to 80°C
0.0006 to 6400 mg/L
2 to 6
Selective
Electrode