missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SI Analytics™ ScienceLine Ion Selective Electrode
SI Analytics ScienceLine Ion Selective Electrodes possess the advantage of a much lower temperature sensitivity and a metal ion free reference system.
1291.43 € - 1630.86 €
Product Code | Brand | Measurement Range | Internal Reference | pH Range | Temperature (Metric) Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Measurement Range | Internal Reference | pH Range | Temperature (Metric) Range | Price | Quantity & Availability | ||
10516063
|
SI Analytics™
285130410 |
0.006 to 127,000 mg/L | Iodide | 0 to 14 | 0°C to 80°C | N/A | |||
10129532
|
SI Analytics™
285130350 |
2 to 35,000 mg/L | Chloride | 2 to 12 | 0°C to 80°C | N/A | |||
10405063
|
SI Analytics™
285130420 |
0.4 to 79,000 mg/L | Bromide | 1 to 12 | 0°C to 80°C | N/A | |||
10405253
|
SI Analytics™
285130430 |
0.0006 to 6400 mg/L | Copper | 2 to 6 | 0°C to 80°C | N/A | |||
10765493
|
SI Analytics™
285130370 |
0.04 to 39,000 mg/L | Potassium | 2 to 12 | 0°C to 40°C | N/A | |||
10211503
|
SI Analytics™
285130400 |
0.003 to 32,000 mg/L | Sulfide | 2 to 12 | 0°C to 80°C | N/A | |||
Description
- ScienceLine sensors for ammonia, sodium, oxygen, ion-selective indicator electrodes
- Also measure pH and temperature
Specifications
Selective | |
Electrode |