missing translation for 'onlineSavingsMsg'
Learn More

Scienceware HDPE Biohazard Bags with Sterilization Patch

These HDPE Biohazard Bags feature a Sterilization Patch, an indicator patch that darkens when sterile.

Brand:  Scienceware 06497-30

Product Code. 11705379

  • 88.65 € / Pack of 100
Estimated Shipment: 30-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Precautionary procedures printed in four languages.
  • Meet/exceed ASTM D 1709-85.
  • Autoclavable at 250°F (121°C).
  • These high-molecular weight bags feature an indicator patch that darkens when sterile.
  • Precautionary procedures are printed in English, French, German, and Spanish.
  • Bags are 1.25 mil thick.
  • Optional bag stands are made of aluminum tubing with baked on epoxy coating.
    • The stands fold flat for convenient storage and include a polystyrene safety drip tray that securely supports the bag in the stand.
  • Order stands and plastic ties separately.
Specifications

Specifications

Biohazard Bag
1 gal.
Clear
Polystyrene safety drip tray that securely supports the bag in the stand
0.310 mm
121°C
610 x 305 mm
610 mm
305 mm
Autoclavable
3.7 L
Storage
HDPE
100/Cs.
0.01 in.
24 in.
12 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Scienceware HDPE Biohazard Bags with Sterilization Patch > HDPE Biohazard Bags with Sterilization Patch; Capacity: 3.7 L (1 gal.); Dimensions (L x W): 610 x 305 mm (24 x 12 in.); Material: HDPE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.