Learn More
DWK Life Sciences Screw Cap closed, with seal for safety standard ground sockets
For safe sealing of DURAN™ threaded sleeves after use.
Brand: DWK Life Sciences 286214202
This item is not returnable.
View return policy
Description
With closed screw caps, devices made of DURAN™ safety grinding can be securely closed after use. The PTFE-coated silicone seal provides maximum chemical resistance.
- Secure closure of the safety ground joints prevents unintentional opening of the ground joint
- PTFE-coated silicone gasket for maximum chemical resistance
Specifications
10/Pk. | |
Suitable for sealing security grinding | |
NS 14/23 |