Learn More
DWK Life Sciences Screw Cap closed, with seal for safety standard ground sockets
For safe sealing of DURAN™ threaded sleeves after use.
41.95 € - 168.00 €
Specifications
Quantity 10/Pk.
View More Specs
Products 6
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
15901551
View Documents
DWK Life Sciences
286214202
NS 14/23
41.95 €
Pack of 10
In Stock
to see stock.
Add to basket
15894598
View Documents
DWK Life Sciences
286214305
NS 19/26
44.90 €
Pack of 10
In Stock
to see stock.
Add to basket
15824608
View Documents
DWK Life Sciences
286214605
NS 34/35
110.00 €
Pack of 10
In Stock
to see stock.
Add to basket
15804608
View Documents
DWK Life Sciences
286214408
NS 24/29
51.70 €
Pack of 10
Estimated Shipment: 25-03-2025
to see stock.
Add to basket
15814608
View Documents
DWK Life Sciences
286214502
NS 29/32
72.90 €
Pack of 10
In Stock
to see stock.
Add to basket
15834608
View Documents
DWK Life Sciences
286214708
NS 45/40
168.00 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
Description
Description
With closed screw caps, devices made of DURAN™ safety grinding can be securely closed after use. The PTFE-coated silicone seal provides maximum chemical resistance.
  • Secure closure of the safety ground joints prevents unintentional opening of the ground joint
  • PTFE-coated silicone gasket for maximum chemical resistance
Specifications
Specifications
10/Pk.