Learn More
Gosselin™ Screw Cap Dippers with Removable Handle
Dip with confidence. Gosselin™ Screw Cap Dippers with Removable Handle prevent spillage with screw caps and detachable shaft.
Brand: Gosselin™ TAT52C-01
Description
- Polypropylene blue containers
- Removable handle
- Shock resistant
- Packaged individually in tamper evident single zip bags
- Traceability information on each bag
- Sterile SAL 10-3
Specifications
Polypropylene | |
Sterile SAL 10-3 | |
52 mm | |
275 mm including container |
125 mL | |
Blue | |
74 mm |