missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
34.30 € - 58.10 €
Chemical Identifiers
CAS | 151-21-3, 7732-18-5 |
---|---|
Molecular Formula | C12H25NaO4S |
Molecular Weight (g/mol) | 288.38 |
MDL Number | MFCD00036175 |
InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
Synonym | Sodium lauryl sulfate, SDS |
PubChem CID | 3423265 |
ChEBI | CHEBI:8984 |
SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10607633
|
Fisher BioReagents
BP1311-200 |
200 mL | Poly Bottle |
34.30 €
200mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10607443
|
Fisher BioReagents
BP1311-1 |
1 L | Nalgene Poly Bottle |
58.10 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis
- DNase, RNase, and protease-free premixed solution containing 20% SDS for protein extraction and DNA extraction procedures
- Reduce preparation time by using this premade SDS solution
- SDS is a commonly used detergent in protein purification, polyacrylamide gel electrophoresis (PAGE), and other molecular biology applications
- SDS is also used to disrupt cell membranes and dissociate nucleic acid-protein complexes in DNA extraction procedures
- Intended for research use only; not for human or animal diagnostic purposes
Chemical Identifiers
151-21-3, 7732-18-5 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
3423265 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
C12H25NaO4S | |
MFCD00036175 | |
Sodium lauryl sulfate, SDS | |
CHEBI:8984 |
Specifications
0.4 max. (0.1% solution) at 230nm, 0.1 max. (0.1% solution) at 280nm | |
151-21-3, 7732-18-5 | |
100°C | |
19.5 to 20.5% | |
(as Fatty Alcohol Sulfate): ≥99% | |
CH3(CH2)10CH2OSO3Na | |
15, 8768 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium;dodecyl sulfate | |
3423265 | |
288.38 | |
DNase-, RNase- and Protease-Free |
Colorless | |
Liquid | |
1.01g/cm³ | |
Pass Test | |
C12H25NaO4S | |
MFCD00036175 | |
Sodium lauryl sulfate, SDS | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
CHEBI:8984 | |
≥99% | |
Sodium Dodecyl Sulfate, 20% Solution |
Safety and Handling
Recommended Storage : RT
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium dodecyl sulfate, 10% solution
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.