Learn More
Cytiva SE260/SE250 MiniVertical Electrophoresis System Replacement Parts: Spineless Combs
Brand:  Cytiva SE211A-10-1.0
93.08 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
 View more versions of this product
Product Code. 10437694
  • 100.82 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit
PTFE
0.03 in.
4.8μL/ mm
0.03 in.
Spineless Comb
10
0.1 cm
0.48 cm
0.1 cm