Learn More
3M™ SecureFit™ X5500 Safety Helmet, Vented, CE
3M™ SecureFit™ Safety Helmet with adjustable positioning and easy-to-adjust, 4-point ratchet suspension system that features 3M ™ Pressure Diffusion Technology. 3M™ UVicator™ Sensor indicates over-exposed to UV-radiation.
Brand: 3M™ 7100175594
217.12 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Secure, comfortable head protection against small falling objects and for general industrial use
- Suspension system incorporates exclusive patented 3M™ Pressure Diffusion Technology
- 4-point ratchet suspension allows wearer to customise height and fit with multiple levels of vertical adjustment
- Vents allow airflow through the helmet
- Optional vents allow airflow through the helmet
- Inspired by climbing helmets with a brimless design to give you a clear upward view
- 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV-radiation and it's time to replace it
- 3M™ Scotchlite™ Reflective Material can be added to increase visibility under low light conditions
- 3M™ Scotchlite™ Reflective Material can also be added to increase visibility under low light conditions
- Approved for lateral deformation (LD)
- Approved for lateral deformation
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Green | |
ABS | |
4-point Ratchet |
Construction, Forestry, Oil, and Gas | |
Safety Helmet | |
Ventilated |