Learn More
Selenium AAS Standard, Fluka™
Certified Reference Material, 1000 mg/L Se in HNO3 2%
Brand:  Honeywell-Fluka E4067-500ML
 View more versions of this product
Product Code. 16676232
  • 55.58 € / 500mL
Explore more special offers
Description
Description
Honeywell Fluka™ analytical standards offers a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. Our products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Selenium AAS Standard
Plastic Bottle
AAS
70°C (Decomposition)
7783-00-8
500 mL
<2
684.9°C