Learn More
Selenium AAS Standard, Fluka™
Certified Reference Material, 1000 mg/L Se in HNO3 2%
33.02 € - 55.58 €
Specifications
Chemical Name or Material | Selenium AAS Standard |
---|---|
CAS | 7783-00-8 |
Packaging | Plastic Bottle |
Grade | AAS |
pH | <2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16666232
|
Honeywell-Fluka
E4067-100ML |
100 mL | N/A | ||||||
16676232
|
Honeywell-Fluka
E4067-500ML |
500 mL |
55.58 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Honeywell Fluka™ analytical standards offers a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. Our products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Selenium AAS Standard | |
Plastic Bottle | |
<2 | |
684.9°C |
7783-00-8 | |
AAS | |
70°C (Decomposition) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.