Learn More
Brady™ Self-laminating Polyester Labels for M710
Printer Labels for use with Brady™ Printer M710.
Brand:  Brady™ 311983
 View more versions of this product
Product Code. 17808034
  • 114.00 € / Pack of 250
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
Specifications
Specifications
ASTM D 1000
Permanent Acrylic
White, Transparent
Label
Single-side Printable
Self-laminating Tag
Laboratory Labeling
1 Label
250 Labels
-321°F to +230°F
0.0019 in.
25.4 x 25.4 mm
1 in.
25.4 mm
0.38 in.
M7-19-461
None
Matte
Polyester
Thermal Transfer
Non-sterile
M710, BMP71
-196°C
Square
-196°C to +110°C
0.048 mm
1 in.
25.4 mm
25.4 mm
9.53 mm