Learn More
Brady™ Self-laminating Polyester Labels for M710
Printer Labels for use with Brady™ Printer M710.
Brand:  Brady™ 311993
 View more versions of this product
Product Code. 17808094
  • 42.95 € / Pack of 250
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
Specifications
Specifications
ASTM D 1000, ASTM D 2979
Permanent Acrylic
White
Label
Single-side Printable
Self-laminating Tag
Healthcare and Hospital Labeling, General Identification, Laboratory Labeling
1 Label
250 Labels
-112°F to +266°F
0.0033 in.
22.86 x 22.86 mm
0.9 in.
22.86 mm
M7-97-481
None
Matte
Polyester
Thermal Transfer
Non-sterile
M710, BMP71
-80°C
Square
-80°C to +130°C
0.084 mm
0.9 in.
22.86 mm