Learn More
Brady™ Self-laminating Polyester Labels for M710
Printer Labels for use with Brady™ Printer M710.
Brand:  Brady™ 311974
 View more versions of this product
Product Code. 17837984
  • 107.00 € / Pack of 250
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
Specifications
Specifications
ASTM D 1000
Permanent Acrylic
White, Transparent
Label
Single-side Printable
Self-laminating Tag
Laboratory Labeling
1 Label
250 Labels
-321°F to +230°F
0.0019 in.
15 x 40 mm
1.57 in.
40 mm
0.59 in.
0.375 in.
M7-101-461
None
Matte
Polyester
Thermal Transfer
Non-sterile
M710, BMP71
-196°C
Rectangle with Circle
-196°C to +110°C
0.048 mm
0.59 in.
40 mm
29.97 mm
15 mm
9.53 mm