Learn More
Brady™ Self-laminating Polyester Labels for M710
Printer Labels for use with Brady™ Printer M710.
Brand:  Brady™ 311977
 View more versions of this product
Product Code. 17848064
  • 66.65 € / Pack of 150
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
Specifications
Specifications
ASTM D 1000
Permanent Acrylic
White, Transparent
Label
Single-side Printable
Self-laminating Tag
Laboratory Labeling
1 Label
150 Labels
-321°F to +230°F
0.0019 in.
41.28 x 15.24 mm
0.6 in.
15.24 mm
1.18 in.
0.5 in.
M7-116-461
None
Matte
Polyester
Thermal Transfer
Non-sterile
M710, BMP71
-196°C
Rectangle with Circle
-196°C to +110°C
0.048 mm
1.625 in.
15.24 mm
15.24 mm
30 mm
12.7 mm