Learn More
Brady™ Self-laminating Polyester Labels for M710
Printer Labels for use with Brady™ Printer M710.
Brand:  Brady™ 311988
 View more versions of this product
Product Code. 17888014
  • 282.00 € / Pack of 250
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
Specifications
Specifications
ASTM D 1000
Permanent Acrylic
White, Transparent
Label
Single-side Printable
Self-laminating Tag
Laboratory Labeling
1 Label
250 Labels
-321°F to +230°F
0.0019 in.
45.72 x 9.53 mm
0.38 in.
9.53 mm
0.75 in.
M7-73-461
None
Matte
Polyester
Thermal Transfer
Non-sterile
M710, BMP71
-196°C
Rectangle
-196°C to +110°C
0.048 mm
1.8 in.
9.53 mm
9.53 mm
19.05 mm