Learn More
Brady™ Self-laminating White Vinyl Wrap Around Wire and Cable Label Tape for M6/M7 Printers
Self-laminating vinyl material is ideal for wire and cable identification. Designed with a clear protective layer to cover the printed text, B-427 material provides superb protection from abrasion, dirt, oils, solvents, water and excess handling.
38.25 € - 84.65 €
Specifications
Autoclavable | Not Autoclavable |
---|---|
Certifications/Compliance | ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized |
Adhesive Type | Permanent Acrylic |
Coating | None |
Color | Clear, White |
Product Code | Brand | Dimensions (LxW) | For Use With (Equipment) | Format | Height (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | For Use With (Equipment) | Format | Height (Metric) | Price | Quantity & Availability | ||
17879194
|
Brady™
174532 |
0.75 x 0.25 in. | M710, BMP71 | Die-cut | 19.05 mm |
48.70 €
Pack of 750 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17878724
|
Brady™
173429 |
0.75 x 0.5 in. | M710, BMP71 | Die-cut | 19.05 mm |
52.10 €
Pack of 500 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17878274
|
Brady™
174180 |
0.75 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 19.05 mm |
55.70 €
Pack of 500 |
In Stock
Log in to see stock. |
||
17878874
|
Brady™
174660 |
0.75 x 1 in. | M710, BMP71 | Die-cut | 19.05 mm |
44.80 €
Pack of 250 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17818794
|
Brady™
173403 |
1 x 0.75 in. | M710, BMP71 | Die-cut | 25.4 mm |
45.40 €
Pack of 250 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17898194
|
Brady™
174216 |
1 x 0.75 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 25.4 mm |
48.50 €
Pack of 250 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17829144
|
Brady™
173353 |
1.5 x 0.75 in. | M710, BMP71 | Die-cut | 38.1 mm |
50.70 €
Pack of 250 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17809204
|
Brady™
174267 |
1.5 x 0.75 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 38.1 mm |
54.25 €
Pack of 250 |
In Stock
Log in to see stock. |
||
17838474
|
Brady™
174208 |
1 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 25.4 mm |
59.30 €
Pack of 500 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17889044
|
Brady™
173411 |
1 x 0.5 in. | M710, BMP71 | Die-cut | 25.4 mm |
55.40 €
Pack of 500 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17818904
|
Brady™
173399 |
1 x 1 in. | M710, BMP71 | Die-cut | 25.4 mm |
49.95 €
Pack of 250 |
In Stock
Log in to see stock. |
||
17878464
|
Brady™
174221 |
1 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 25.4 mm |
66.65 €
Pack of 250 |
In Stock
Log in to see stock. |
||
17898744
|
Brady™
174533 |
1 x 1.5 in. | M710, BMP71 | Die-cut | 25.4 mm |
80.25 €
Pack of 250 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17848784
|
Brady™
174593 |
1.5 x 0.25 in. | M710, BMP71 | Die-cut | 38.1 mm |
62.40 €
Pack of 750 |
In Stock
Log in to see stock. |
||
17828864
|
Brady™
173362 |
1.5 x 0.5 in. | M710, BMP71 | Die-cut | 38.1 mm |
67.95 €
Pack of 500 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17899014
|
Brady™
173345 |
1.5 x 1 in. | M710, BMP71 | Die-cut | 38.1 mm |
64.25 €
Pack of 250 |
In Stock
Log in to see stock. |
||
17888894
|
Brady™
173337 |
1.5 x 1.5 in. | M710, BMP71 | Die-cut | 38.1 mm |
65.85 €
Pack of 250 |
In Stock
Log in to see stock. |
||
17878754
|
Brady™
174651 |
1.5 x 1.75 in. | M710, BMP71 | Die-cut | 38.1 mm |
76.85 €
Pack of 250 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17889194
|
Brady™
174534 |
1.5 x 2 in. | M710, BMP71 | Die-cut | 38.1 mm |
84.65 €
Pack of 100 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17828734
|
Brady™
174531 |
1.25 x 1 in. | M710, BMP71 | Die-cut | 31.75 mm |
55.40 €
Pack of 250 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17868334
|
Brady™
174275 |
1.5 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 38.1 mm |
68.70 €
Pack of 250 |
In Stock
Log in to see stock. |
||
17838404
|
Brady™
174283 |
1.5 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 38.1 mm |
70.40 €
Pack of 250 |
In Stock
Log in to see stock. |
||
17818404
|
Brady™
174257 |
1.5 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 38.1 mm |
72.65 €
Pack of 500 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17808304
|
Brady™
174236 |
2.5 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 63.5 mm |
51.05 €
Pack of 100 |
In Stock
Log in to see stock. |
||
17889204
|
Brady™
173378 |
2.5 x 1 in. | M710, BMP71 | Die-cut | 63.5 mm |
38.25 €
Pack of 100 |
In Stock
Log in to see stock. |
||
17829164
|
Brady™
174640 |
3.15 x 1.938 in. | M710, BMP71 | Die-cut | 80 mm |
64.80 €
Pack of 100 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17818224
|
Brady™
173316 |
3.375 x 1 in. | M710, BMP71 | Die-cut | 85.73 mm |
42.60 €
Pack of 100 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17818444
|
Brady™
173894 |
3.15 x 1.938 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 80.01 mm |
69.30 €
Pack of 100 |
In Stock
Log in to see stock. |
||
17838304
|
Brady™
174338 |
3.375 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 85.73 mm |
39.70 €
Pack of 100 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17868214
|
Brady™
174243 |
4 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 101.6 mm |
48.75 €
Pack of 100 |
In Stock
Log in to see stock. |
||
17848124
|
Brady™
174285 |
4 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 101.6 mm |
66.00 €
Pack of 100 |
In Stock
Log in to see stock. |
||
17819024
|
Brady™
174535 |
4 x 1.5 in. | M710, BMP71 | Die-cut | 101.6 mm |
56.75 €
Pack of 100 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17858894
|
Brady™
173335 |
4 x 1.5 in. | M710, BMP71 | Die-cut | 101.6 mm |
61.65 €
Pack of 100 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
17858984
|
Brady™
173372 |
4 x 1 in. | M710, BMP71 | Die-cut | 101.6 mm |
45.65 €
Pack of 100 |
In Stock
Log in to see stock. |
||
17829174
|
Brady™
173332 |
6 x 1.5 in. | M710, BMP71 | Die-cut | 152.4 mm |
51.20 €
Pack of 50 |
In Stock
Log in to see stock. |
||
17858214
|
Brady™
174288 |
6 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 152.4 mm |
54.75 €
Pack of 50 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||
Description
- Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text
- Durable, permanent acrylic adhesive has outstanding water and oil resistance
- UL approved
- Service temperature of -40°C to +70°C (-40°F to +158°F)
- Made of B-427 vinyl material
Specifications
Not Autoclavable | |
Permanent Acrylic | |
Clear, White | |
Vinyl | |
Thermal Transfer | |
Non-sterile | |
No | |
-70°C to +70°C | |
0.102 mm |
ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | |
None | |
Matte | |
Single-side Printable | |
Labels | |
Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling | |
-94°F to +158°F | |
0.004 in. |