Learn More
Keystone Cleanroom Products Self Seal Tyvek™ Steam Sterilization Pouches
Self Seal Tyvek™ Steam Sterilization Pouches provide an excellent barrier from microbial contamination.
Brand:  Keystone Cleanroom Products 01626TWBBSSSDSP02
 View more versions of this product
Product Code. 17841711
  • 3610.00 € / Pack of 500
Explore more special offers
Description
Description
  • Great choice for equipment parts that are heavy, have sharp edges, or packaging that requires surface sanitization post-sterilization
  • DuPont Tyvek™/Polyester-based flexible film is lint-free, moisture and puncture resistant, extremely tough, and designed to be autoclaved
  • The Tyvek™ side offers high breathability while providing an excellent barrier from microbial contamination
  • The clear film side offers excellent clarity along with strength and durability
  • These bags feature a medical grade adhesive strip for convenient self-sealing
  • Each pouch has a CLASS A process indicator, which changes from pink to dark brown when exposed to steam
  • When the peel-able seals are opened, virtually no airborne particulates are generated
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Eliminates the need for the use and validation of a Heat Sealer
Specifications
Specifications
Autoclavable
Self-seal
Steam Sterilization
HDPE
16 x 26 in.
40.6 cm
66 cm
Certificate of Conformance (COC)
White
Steam Indicator Dot
25/Bag, 500/Cs.
16 in.
26 in.