Learn More
BD Sentry™ Safety Lancet
Allows for quick and safe blood collection from the fingertip
Brand:  BD 369523
Product Code. 16611592
  • 219.00 € / Pack of 100
Edge
Explore more special offers
Description
Description
  • Activated by the push of a button
  • Mechanism to prevent reuse and needlestick injuries
  • Easy disposal in sharps containers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Green
Single-use
100/Pk.
1.8 mm
23G
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Sentry™ Safety Lancet > Sterile, 23G, 1.8 mm, Green, 100/Pk.