Learn More
Cytiva Sera-Mag™ Carboxylate modified Magnetic Beads, hydrophobic
Excellent for coupling oligos or proteins directly to beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these single shell magnetite beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.
Brand:  Cytiva 44152105050450
 View more versions of this product
Product Code. 11889782
Description
Description
  • High binding capacity
  • 1.0μm diameter
  • High carboxyl acid content for binding for binding DNA and proteins
  • Hydrophobic bead based on crosslinking requirements for the encapsulation layer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5% solids (50mg/mL)
DNA Sample Prep, Protein Purification
1 μm
2°C to 8°C
1 L