missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag™ Carboxylate modified Magnetic Beads, hydrophobic
Excellent for coupling oligos or proteins directly to beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these single shell magnetite beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.
405.00 € - 10500.00 €
Specifications
Concentration | 5% solids (50mg/mL) |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | DNA Sample Prep, Protein Purification |
Diameter (Metric) | 1 μm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13273589
|
Cytiva
44152105050250 |
15 mL |
405.00 €
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
11879782
|
Cytiva
44152105050350 |
100 mL |
2147.00 €
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
11889782
|
Cytiva
44152105050450 |
1 L | |||||||
Description
- High binding capacity
- 1.0μm diameter
- High carboxyl acid content for binding for binding DNA and proteins
- Hydrophobic bead based on crosslinking requirements for the encapsulation layer
Specifications
5% solids (50mg/mL) | |
2°C to 8°C | |
DNA Sample Prep, Protein Purification | |
1 μm |