Learn More
Cytiva Sera-Mag SpeedBeads Streptavidin-Blocked Magnetic Particles
Combines high affinity for biotinylated target molecules with very low nonspecific binding.
Brand:  Cytiva 21152104010150
787.66 EUR valid until 2024-12-31
Use promo code "21948" to get your promotional price.
 View more versions of this product
Product Code. 11894972
  • 924.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Combines fast reaction kinetics with very low non-specific binding for increased throughput and precision
  • Applications include automated immunoassays, immunoprecipitation, and protein purification
  • Streptavidin coating targets Biotinylated Molecules
  • Compounds that are difficult to attach to particle surfaces by conventional means may be amenable to biotinylation
  • Provides a high biotin-binding capacity along with a strong affinity for targeted, biotin labeled molecules
  • The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer
  • Two layers of magnetite are coated onto this core, resulting in faster magnetic response times
  • The surface is blocked with a non-surfactant, non-protein based method, to help prevent non-specific binding of proteins
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    10 mg/mL
    5 mL
    1 μm