missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag SpeedBeads Streptavidin-Blocked Magnetic Particles

Combines high affinity for biotinylated target molecules with very low nonspecific binding.

403.00 € - 924.00 €

Specifications

Concentration 10 mg/mL
Diameter (Metric) 1 μm
View More Specs

Products 2
Product Code Brand Packaging Price Quantity & Availability  
Product Code Brand Packaging Price Quantity & Availability  
11814982
View Documents
Cytiva
21152104011150
1 mL
403.00 €
Each
In Stock
to see stock.
Add to basket
 
11894972
View Documents
Cytiva
21152104010150
5 mL
924.00 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • Combines fast reaction kinetics with very low non-specific binding for increased throughput and precision
  • Applications include automated immunoassays, immunoprecipitation, and protein purification
  • Streptavidin coating targets Biotinylated Molecules
  • Compounds that are difficult to attach to particle surfaces by conventional means may be amenable to biotinylation
  • Provides a high biotin-binding capacity along with a strong affinity for targeted, biotin labeled molecules
  • The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer
  • Two layers of magnetite are coated onto this core, resulting in faster magnetic response times
  • The surface is blocked with a non-surfactant, non-protein based method, to help prevent non-specific binding of proteins
  • Specifications

    Specifications

    10 mg/mL
    1 μm
    Videos
    SDS
    Documents

    Documents

    Product Certifications