Learn More
Cytiva Sera-Mag SpeedBeads™ Streptavidin magnetic beads
SpeedBeads Streptavidin exhibit a high affinity when binding to biotinylated ligands such as proteins, nucleic acids and peptides. They respond much faster to a magnetic field to shorten assay times and improve precision and also move faster through viscous solutions.
Brand:  Cytiva 66152104010150
826.20 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
 View more versions of this product
Product Code. 11849912
  • 935.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available with a 3500-4500 pmol/mg biotin-binding range
  • Their cauliflower-like structure gives Sera-Mag beads high binding capacity
  • Combine fast reaction kinetics and low, non-specific binding for increased throughput and precision in such immunoassay and molecular biology applications as sample preparation and assay development for genomics and proteomics
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1% solids (10mg/mL)
Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets
1 μm
2°C to 8°C
5 mL