missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag SpeedBeads™ Streptavidin magnetic beads
SpeedBeads Streptavidin exhibit a high affinity when binding to biotinylated ligands such as proteins, nucleic acids and peptides. They respond much faster to a magnetic field to shorten assay times and improve precision and also move faster through viscous solutions.
294.00 € - 5259.00 €
Specifications
Concentration | 1% solids (10mg/mL) |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets |
Diameter (Metric) | 1 μm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11869912
|
Cytiva
66152104011150 |
1 mL |
294.00 €
Each |
Estimated Shipment: 04-03-2025
Log in to see stock. |
|||||
11849912
|
Cytiva
66152104010150 |
5 mL |
935.00 €
Each |
In Stock
Log in to see stock. |
|||||
11859912
|
Cytiva
66152104010350 |
100 mL |
5259.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- Available with a 3500-4500 pmol/mg biotin-binding range
- Their “cauliflower-like” structure gives Sera-Mag beads high binding capacity
- Combine fast reaction kinetics and low, non-specific binding for increased throughput and precision in such immunoassay and molecular biology applications as sample preparation and assay development for genomics and proteomics
Specifications
1% solids (10mg/mL) | |
2°C to 8°C | |
Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets | |
1 μm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title