Learn More
Cytiva Sera-Mag SpeedBeads™ Streptavidin magnetic beads
SpeedBeads Streptavidin exhibit a high affinity when binding to biotinylated ligands such as proteins, nucleic acids and peptides. They respond much faster to a magnetic field to shorten assay times and improve precision and also move faster through viscous solutions.
Marke:  Cytiva 66152104010350
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 11859912
  • 5259.00 € / Each
In Stock
to see stock.
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Beschreibung
Beschreibung
  • Available with a 3500-4500 pmol/mg biotin-binding range
  • Their cauliflower-like structure gives Sera-Mag beads high binding capacity
  • Combine fast reaction kinetics and low, non-specific binding for increased throughput and precision in such immunoassay and molecular biology applications as sample preparation and assay development for genomics and proteomics
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
1% solids (10mg/mL)
Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets
1 μm
2°C to 8°C
100 mL
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote