Learn More
Cytiva Sera-Mag SpeedBeads™ Streptavidin magnetic beads
SpeedBeads Streptavidin exhibit a high affinity when binding to biotinylated ligands such as proteins, nucleic acids and peptides. They respond much faster to a magnetic field to shorten assay times and improve precision and also move faster through viscous solutions.
Brand: Cytiva 66152104011150
266.64 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Available with a 3500-4500 pmol/mg biotin-binding range
- Their cauliflower-like structure gives Sera-Mag beads high binding capacity
- Combine fast reaction kinetics and low, non-specific binding for increased throughput and precision in such immunoassay and molecular biology applications as sample preparation and assay development for genomics and proteomics
Especificaciones
1% solids (10mg/mL) | |
Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets | |
1 μm |
2°C to 8°C | |
1 mL |