missing translation for 'onlineSavingsMsg'
Learn More

Cytiva SeraSil-Mag Silica Coated Superparamagnetic Beads

SeraSil-Mag beads for nucleic acid isolation deliver high purity DNA extraction in NGS applications.

Brand:  Cytiva 29357374

 View more versions of this product

Product Code. 16467655

  • 474.00 € / 60mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Delivers high purity DNA extraction for highly sensitive applications where sample is scarce
  • These beads provide an optimal surface for nucleic acid binding with high performance and low background
  • High magnetization (60 emu/g) and strong binding capacity giving fast magnetic response (∼5 secs) and shorten time required for magnetic steps during isolation
  • Beads come in two sizes (submicroscale diameter 700 nm and 400 nm), offering choice based on surface requirements and high binding capacity due to the small sized particles
  • Beads are monodispersed with narrow size distribution for consistent, reproducible results
  • Silanol hydroxyl groups on the bead surface give efficient, high purity isolation of nucleic acids
  • Tested for microbial contamination
  • Good buoyancy stability (low sedimentation rate): beads remain well suspended and dispersed for over 90 mins (at 1 mg/mL concentration)
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    20 mg/mL in water (0.05% sodium azide)
    Storage Solution: Water + 0.05% sodium azide
    Chaotropic salt chemistry and vortex mixing, roller mixing and sonication
    60 mL
    Room Temperature
    Room Temperature
    700 nm
    Silica coated, core-shell structure
    700 nm diameter particle
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Cytiva SeraSil-Mag Silica Coated Superparamagnetic Beads > Quantity: 60 mL, dimParticleSizeMetric: 700 nm diameter

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.