missing translation for 'onlineSavingsMsg'
Learn More

3M™ Series G3000 Ventilated Slim Design Hard Hat with Leather Sweatband

Series G3000 Ventilated Slim Design Hard Hat with Leather Sweatband allows falling objects to slide off easily and not dislodge the helmet from its position.

Brand:  3M™ 7000108296

380.92 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15749977

  • 471.00 € / Pack of 20
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • A hard hat designed for use in harsh environments, with excellent ventilation and a good field of vision.
  • 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV- radiation and its time to replace it.
  • Softly rounded design, which helps to avoid helmet parts snagging on branches, etc.
  • Ventilation holes distributed over a larger area offer maximum through-flow of air increasing wearer comfort.
  • Head harness can be rotated 180° for better visibility when working in confined spaces.
  • Short peak gives improved upwards visibility.
  • Easy to replace soft leather sweatband for the highest level of comfort and hygiene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EN 397
UV-stabilized ABS
54 to 62 cm
Ventilated
Green
Helmet
310 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.