Learn More
Falcon™ Serological Pipette, Triple Bagged
Corning’s triple-bagged packaged pipets are a safety and convenience revolution. This packaging protects the product from external contamination and is ideal for laboratory clean rooms, such as cell-based vaccine or drug production facilities.
Brand:  Falcon™ 355028
 View more versions of this product
Product Code. 30093042
  • 218.00 € / Pack of 200
Estimated Shipment: 28-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Triple-bagged in two outer bags
  • Manufactured and packaged with animal-free materials
  • Sterility Assurance Level (SAL) of 10-6
  • USP Class VI tested
  • Non-pyrogenic
  • Non-cytotoxic
  • RNase-/DNase-free
  • Human DNA-free
  • Low particulate according to USP<788>
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plastic
Yes
Triple Bagged, Bulk
25 mL
Reusable
Sterile
200/Pk.
Transparent
Serological Pipet
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Falcon™ Serological Pipette, Triple Bagged > Triple Bag Serological Pipette; Size: 25 mL
Unless otherwise specified, all products are for research use or general laboratory use only. Not intended for use in diagnostic or therapeutic procedures. Not for use in humans. These products are not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment. Corning Life Sciences makes no claims regarding the performance of these products for clinical or diagnostic applications.
Corning's products are not specifically designed and tested for diagnostic testing. Many Corning products, though not specific for diagnostic testing, can be used in the workflow and preparation of the test at the customers discretion. Customers may use these products to support their claims. We cannot make any claims or statements that our products are approved for diagnostic testing either directly or indirectly. The customer is responsible for any testing, validation, and/or regulatory submissions that may be required to support the safety and efficacy of their intended application.