missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Diamond CTC PAL 10 to 50μL GC Autosampler Syringes

Improves sensitivity of MS Analysis with reduced non specific sample interaction and increased analyte recovery.

Brand:  Trajan™

 View more versions of this product

Product Code. 13107946

  • 96.45 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Ensures carryover is reduced to levels lower even than the best fixed needle syringes available. In the event of needle damage, Diamond MS needles provide a cost effective alternative to purchasing an entire fixed needle syringe.
  • Ultra Smooth needles reduce non specific interaction between your sample and the needle. 
  • Further reduce sample interactions with a hydrophobic or hydrophillic needle coating
Specifications

Specifications

23 G
0.11 mm
0.63 mm
Ultra Smooth Coating
GC Autosampler Syringe
10 to 50 μL
Cone
0.004 in.
0.024 in.
Gas chromatography
1.96 in.
Ea.
50 mm
CTC PAL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.