Learn More
Trajan™ Diamond CTC PAL 100μL GC Autosampler Syringes
Improves sensitivity of MS Analysis with reduced non specific sample interaction and increased analyte recovery.
Brand: Trajan™ 0355521
This item is not returnable.
View return policy
Description
- Ensures carryover is reduced to levels lower even than the best fixed needle syringes available. In the event of needle damage, Diamond MS needles provide a cost effective alternative to purchasing an entire fixed needle syringe.
- Ultra Smooth needles reduce non specific interaction between your sample and the needle.
- Further reduce sample interactions with a hydrophobic or hydrophillic needle coating
Specifications
23 G | |
0.24 mm | |
0.63 mm | |
1.9 in. | |
1 Pk. | |
Ultra Smooth Hydrophobic, Cone |
0.009 in. | |
0.024 in. | |
Gas chromatography | |
Autosampler Syringe | |
100 μL | |
CTC PAL GC |