Learn More
Trajan™ Diamond CTC PAL 100μL GC Autosampler Syringes
Improves sensitivity of MS Analysis with reduced non specific sample interaction and increased analyte recovery.
Brand:  Trajan™ 0355531
 View more versions of this product
Product Code. 13197936
  • 96.45 € / Each
Explore more special offers
Description
Description
  • Ensures carryover is reduced to levels lower even than the best fixed needle syringes available. In the event of needle damage, Diamond MS needles provide a cost effective alternative to purchasing an entire fixed needle syringe.
  • Ultra Smooth needles reduce non specific interaction between your sample and the needle. 
  • Further reduce sample interactions with a hydrophobic or hydrophillic needle coating
Specifications
Specifications
23 G
0.24 mm
0.63 mm
1.9 in.
1 Pk.
Ultra Smooth, Cone
0.009 in.
0.024 in.
Gas chromatography
Autosampler Syringe
100 μL
CTC PAL GC