missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ BP21 (FFAP) GC Capillary Columns
A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids
Brand: Trajan™ 054474
This item is not returnable.
View return policy
Description
A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids
- Nitroterephthalic acid modified PEG polar phase
- Ideal for low molecular weight acids
- Applications include volatile free acids, fatty acid methyl esters (FAMEs), alcohols, aldehydes and ketones
Specifications
250°C | |
0.53 mm | |
G25, G35 | |
35°C to 240/250°C | |
25 m | |
Polar |
Hydrogen Bonding or Dipole interactions | |
Polyethylene Glycol | |
FAMEs Analysis, Fatty Acid Methyl Esters, Alcohol Analysis, Aldehydes Analysis, Acrylates, Ketones | |
0.5 μm | |
BP21 (FFAP) | |
Fused Silica |