missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ BP21 (FFAP) GC Capillary Columns

A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids

720.00 € - 1340.00 €

Specifications

Stationary Phase Polyethylene Glycol
USP Type G25, G35
Temperature Range 35°C to 240/250°C
Packing Material BP21 (FFAP)
Polarity Polar
View More Specs

Products 10
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
12945591
View Documents Promotion Details
Trajan™
054462
25 m 0.22 mm 0.25 μm
720.00 €
Each
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
12985591
View Documents Promotion Details
Trajan™
054469
50 m 0.32 mm 0.25 μm
1340.00 €
Each
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
12905601
View Documents Promotion Details
Trajan™
054474
25 m 0.53 mm 0.5 μm
850.00 €
Each
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
12915601
View Documents Promotion Details
Trajan™
054465
30 m 0.25 mm 0.25 μm
720.00 €
Each
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
11967235
View Documents Promotion Details
Trajan™
054466
60 m 0.25 mm 0.25 μm
1150.00 €
Each
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
15268683
View Documents Promotion Details
Trajan™
054471
30 m 0.32 mm 0.25 μm
770.00 €
Each
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
15278683
View Documents Promotion Details
Trajan™
054472
60 m 0.32 mm 0.25 μm
1340.00 €
Each
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
12935601
View Documents Promotion Details
Trajan™
054477
30 m 0.53 mm 0.5 μm
850.00 €
Each
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
12975591
View Documents Promotion Details
Trajan™
054468
25 m 0.32 mm 0.25 μm
770.00 €
Each
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
15288683
View Documents Promotion Details
Trajan™
054478
30 m 0.53 mm 1 μm
850.00 €
Each
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
Description

Description

A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids

  • Nitroterephthalic acid modified PEG polar phase
  • Ideal for low molecular weight acids
  • Applications include volatile free acids, fatty acid methyl esters (FAMEs), alcohols, aldehydes and ketones
Specifications

Specifications

Polyethylene Glycol
35°C to 240/250°C
Polar
G25, G35
BP21 (FFAP)
Fused Silica
Videos
SDS
Documents

Documents

Product Certifications