missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ BP21 (FFAP) GC Capillary Columns

A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids

Brand:  Trajan™ 054477

 View more versions of this product

Product Code. 12935601

  • 850.00 € / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids

  • Nitroterephthalic acid modified PEG polar phase
  • Ideal for low molecular weight acids
  • Applications include volatile free acids, fatty acid methyl esters (FAMEs), alcohols, aldehydes and ketones
Specifications

Specifications

250°C
0.53 mm
G25, G35
35°C to 240/250°C
30 m
Polar
Hydrogen Bonding or Dipole interactions
Polyethylene Glycol
Volatile Free Acids Analysis, Fatty Acid Methyl Esters, Alcohol Analysis, Aldehydes Analysis, Acrylates, Ketones
0.5 μm
BP21 (FFAP)
Fused Silica
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers