Learn More
Trajan™ GC Capillary Column Connectors: Replacement Ferrules
Replacement SilTite ferrules for SilTiteμ-Union
Brand:  Trajan™ 073568
 View more versions of this product
Product Code. 15208943
  • 155.00 € / Pack of 10
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • SilTite μ-Union gives low thermal mass - 9mm in length and <0.5g mass
  • Zero dead volume for optimized peak shape
  • Chemically deactivated stainless steel ferrules give system inertness
  • Robust and stable for both high temperature and pressure cycling
Specifications
Specifications
Replacement μ-Union Ferrule
Stainless Steel
0.1 to 0.25 mm I.D. First Column with 0.4 mm Holes and 0.53 mm I.D. Second Column with 0.8 mm Holes